Order Entry
Finland
ContactUsLinkComponent
Anti-GLB1 Recombinant Antibody [Clone: 13R4]
Anti-GLB1 Recombinant Antibody [Clone: 13R4]
Catalog # PRSI24-155
Supplier:  ProSci Inc.
New Product
Anti-GLB1 Recombinant Antibody [Clone: 13R4]
Catalog # PRSI24-155
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    galactosidase beta 1
  • Antigen symbol:
    GLB1
  • Clonality:
    Recombinant
  • Clone:
    13R4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Isotype:
    IgG2c lambda
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    2720
  • Antigen synonyms:
    MPS4B|EBP|β-galactosidase|β-gal|GLB1|beta-gal|Acid beta galactosidase|ELNR1|Galactosidase beta 1|Beta galactosidase 1
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Beta-galactosidase.
  • Tested applications:
    negative control
  • Size:
    0,2 mg
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: galactosidase beta 1
Clonality: Recombinant
Clone: 13R4
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2c lambda
Reactivity: E. coli