Anti-C3 Recombinant Antibody [Clone: 528]
Catalog # PRSI24-137
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:complement C3
- Antigen symbol:C3
- Clonality:Recombinant
- Clone:528
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:718
- Antigen synonyms:PIG61|HER1|epidermal growth f|mENA|proto-oncogene c-ErbB-1|ERBB|avian erythroblastic leukemia viral (v-erb-b) oncogene homolog|cell proliferation-inducing protein 61|cell growth inhibiting protein 40|receptor tyrosine-protein kinase erbB-1|ERBB1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:Partially purified EGFR from A431 cells.
- Tested applications:NTRL
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: complement C3
Clonality: Recombinant
Clone: 528
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG2a kappa
Reactivity: Human