Anti-BAX Recombinant Antibody [Clone: 6A7]
Catalog # PRSI24-115
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:BCL2 associated X, apoptosis regulator
- Antigen symbol:BAX
- Clonality:Recombinant
- Clone:6A7
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Gene ID:581
- Antigen synonyms:Bcl2-L-4|Apoptosis regulator BAX|Bcl-2-like protein 4
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:KLH conjugated to peptides corresponding to amino acids 3–16 of murine Bax (CGSGEQLGSGGPTSS), amino acids 3–16 of human Bax (CGSGEQPRGGGPTSS), and amino acids 12–24 shared by Bax from both species (CGPTSSEQIMKTGA).
- Size:0,2 mg
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: BCL2 associated X, apoptosis regulator
Clonality: Recombinant
Clone: 6A7
Conjugation: Unconjugated
Host: Recombinant
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat