- Antibody type:Primary
- Antigen name:breast cancer 1, early onset
- Antigen symbol:BRCA1
- Clonality:Monoclonal
- Clone:BRCA1/1472
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P38398
- Antigen synonyms:BRCA1
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Amino acids 445-620 were used as the immunogen for the BRCA1 antibody.
- Purification:Protein G affinity chromatography
- Size:100 µg
- Pk:100 µG
BRCA1 (Breast cancer type 1 susceptibility protein) is a nuclear phosphoprotein that plays a role in maintaining genomic stability, and it also acts as a tumor suppressor. The encoded protein combines with other tumor suppressors, DNA damage sensors, and signal transducers to form a large multi-subunit protein complex known as the BRCA1-associated genome surveillance complex (BASC). This gene product associates with RNA polymerase II, and through the C-terminal domain, also interacts with histone deacetylase complexes. This protein thus plays a role in transcription, DNA repair of double-stranded breaks, and recombination. Mutations in this gene are responsible for approximately 40% of inherited breast cancers and more than 80% of inherited breast and ovarian cancers. Alternative splicing plays a role in modulating the subcellular localization and physiological function of this gene.
Flow Cytometry: 0,5-1 µg/million cells in 0,1 ml Immunofluorescence: 1 - 2 µg/ml ELISA: order the BSA free formulation for coating Optimal dilution of the BRCA1 antibody should be determined by the researcher.
Type: Primary
Antigen: BRCA1
Clonality: Monoclonal
Clone: BRCA1/1472
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human