Order Entry
Finland
ContactUsLinkComponent
Anti-TNFSF13B Mouse Monoclonal Antibody [clone: C257/1638]
Anti-TNFSF13B Mouse Monoclonal Antibody [clone: C257/1638]
Catalog # PRSI33-595
Supplier:  ProSci Inc.
New Product
Anti-TNFSF13B Mouse Monoclonal Antibody [clone: C257/1638]
Catalog # PRSI33-595
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tumor necrosis factor (ligand) superfamily, member 13b
  • Antigen symbol:
    TNFSF13B
  • Clonality:
    Monoclonal
  • Clone:
    C257/1638
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Format:
    Purified Antibody
  • Form:
    Liquid
  • Gene ID:
    Q9Y275
  • Antigen synonyms:
    TNFSF13B
  • Storage buffer:
    PBS with 0,1 mg/ml BSA and 0,05% sodium azide
  • Storage temperature:
    Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
  • Concentration:
    0,2 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    Recombinant human BAFF protein was used as the immunogen for this BAFF antibody.
  • Purification:
    Protein G affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

BAFF/BLyS/CD257 is a type II transmembrane protein, and a member of the tumor necrosis factor ligand superfamily. It is proteolytically cleaved to form a soluble protein. BAFF, like other TNF ligand family members, forms homo-trimers. The predicted molecular weight of a BAFF monomer is approximately 31 kDa. It is expressed on peripheral blood B and T lymphocytes, monocytes, macrophages, and dendritic cells and is upregulated by IFN-gamma and down-regulated by PMA/ionomycin treatment. BAFF stimulates B and T cell immunity and regulates humoral immunity by binding TACI and BCMA receptors; BAFF prevents apoptosis and promotes B cell survival by binding the BAFF-specific receptor (BAFFR/BR3).

Flow Cytometry: 0,5-1 µg/million cells IF: 1 - 2 µg/ml Titering of the BAFF antibody may be required for optimal performance.

Type: Primary
Antigen: TNFSF13B
Clonality: Monoclonal
Clone: C257/1638
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human