Specifications
- Antibody type:Primary
- Antigen name:fucosyltransferase 4 (alpha (1,3) fucosyltransferase, myeloid-specific)
- Antigen symbol:FUT4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Format:Purified Antibody
- Form:Liquid
- Gene ID:P22083
- Antigen synonyms:FUT4
- Storage buffer:PBS with 0,1 mg/ml BSA and 0,05% sodium azide
- Storage temperature:Aliquot and Store at 2…8 °C. Avoid freez-thaw cycles.
- Concentration:0,2 mg/ml
- Shipping temperature:4 °C
- Immunogen:Purified human neutrophils were used as the immunogen for the CD15 antibody.
- Purification:Protein A affinity
- Size:100 µg
- Pk:100 µG
Specifications
About this item
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on >95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin s disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.
Flow Cytometry: 2-4 µg/million cells in 0,1 ml Immunofluorescence: 2-4 µg/ml Immunohistochemistry (FFPE): 5-10 µg/ml for 30 min at RT Optimal dilution of the CD15 antibody should be determined by the researcher.
Type: Primary
Antigen: FUT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat