Anti-PSME3 Rabbit Polyclonal Antibody
Catalog # PRSI14-585
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:proteasome (prosome, macropain) activator subunit 3 (PA28 gamma; Ki)
- Antigen symbol:PSME3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:10197
- Antigen synonyms:HEL-S-283|PA28G|PSME3|PA28gamma|REG-GAMMA|PA28-gamma|Ki
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at –20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human PSME3 (NP_005780.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: PSME3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat