Anti-SETD8 Rabbit Polyclonal Antibody
Catalog # PRSI22-850
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:SET domain containing (lysine methyltransferase) 8
- Antigen symbol:SETD8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:387893
- Antigen synonyms:PRSET7|SETD8|SET07|KMT5A|H4-K20-HMTase SETD8|PR/SET07|Lysine N-methyltransferase 5A|SET8|PR-Set7
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 73-352 of human SETD8 (NP_065115.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SETD8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat