Anti-DUSP22 Rabbit Polyclonal Antibody
Catalog # PRSI22-832
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Dual specificity phosphatase 22
- Antigen symbol:DUSP22
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:56940
- Antigen synonyms:epididymis secretory sperm binding protein|dual specificity protein phosphatase 22|homolog of mouse dual specificity phosphatase LMW-DSP2|JNK-stimulatory phosphatase-1|JSP1|LMWDSP2|MKPX|LMW-DSP2|JKAP|MKP-x|JNK-stimulating phosphatase 1|low molecular weight dual specificity phosphatase 2|MAP kinase phosphatase x|JSP-1|VHX|mitogen-activated protein kinase phosphatase x
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-184 of human DUSP22 (NP_064570.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: DUSP22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human