Anti-ABCA3 Rabbit Polyclonal Antibody
PRSI22-511
New Product
- Antibody type:Primary
- Antigen name:ATP binding cassette subfamily A member 3
- Antigen symbol:ABCA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:21
- Antigen synonyms:sub-family A (ABC1)|ABC-C|LBM18SMDP3|EST111653|ABC3|ABC transporter 3|member 3|ABC-C transporter|ATP-binding cassette|ATP-binding cassette sub-family A member 3|ATP-binding cassette transporter 3
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 43-203 of human ABCA3 (NP_001080.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: ABCA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human