Anti-RAD1 Rabbit Polyclonal Antibody
PRSI22-505
New Product
- Antibody type:Primary
- Antigen name:RAD1 homolog (S. pombe)
- Antigen symbol:RAD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Form:Liquid
- Gene ID:5810
- Antigen synonyms:RAD1|DNA repair exonuclease REC1|RAD1 (S. pombe) homolog|Exonuclease homolog RAD1|REC1|HRAD1|RAD1 homolog (S. pombe)
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-282 of human RAD1 (NP_002844.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: RAD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat