Anti-POLR2G Rabbit Polyclonal Antibody
Catalog # PRSI22-504
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) II (DNA directed) polypeptide G
- Antigen symbol:POLR2G
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5436
- Antigen synonyms:POLR2G|DNA directed RNA polymerase II polypeptide G|MGC138369|MGC138367|hsRPB7|RPB7|hRPB19|RNA polymerase II seventh subunit|DNA directed RNA polymerase II 19 kda polypeptide|polymerase (RNA) II (DNA directed) polypeptide G
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human POLR2G (NP_002687.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: POLR2G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat