Anti-POLR2F Rabbit Polyclonal Antibody
PRSI22-503
New Product
- Antibody type:Primary
- Antigen name:RNA polymerase II subunit F
- Antigen symbol:POLR2F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:5435
- Antigen synonyms:RPB14.4|and III 14.4 kDa polypeptide|RNA polymerases I|RPABC2|DNA-directed RNA polymerases I|polymerase (RNA) II subunit F|and III subunit RPABC2|II|RPABC14.4|polymerase (RNA) II (DNA directed) polypeptide F|RNA Polymerase II subunit 14.4 kD|and III subunit ABC2|RPC15|HRBP14.4|POLRF|RPB6|DNA-directed RNA polymerase II subunit F
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human POLR2F (NP_068809.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: POLR2F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat