Anti-AGO2 Rabbit Polyclonal Antibody
PRSI22-494
New Product
- Antibody type:Primary
- Antigen name:Argonaute RISC catalytic component 2
- Antigen symbol:AGO2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:27161
- Antigen synonyms:LINC0098PPD|EIF2C2|cancer susceptibility candidate 7 (non-protein coding)|cancer susceptibility candidate 7|Q1protein argonaute-2|RISC catalytic component|argonaute 2|PAZ Piwi domain protein|long intergenic non-protein coding RNA 98protein slicer|CASC7|2|eukaryotic translation initiation factor 2C
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 47-115 of human EIF2C2 (NP_036286.2)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: AGO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse