Anti-SCYL1 Rabbit Polyclonal Antibody
Catalog # PRSI22-447
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:N-terminal kinase-like protein
- Antigen symbol:SCYL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:57410
- Antigen synonyms:SCYL1|NTKL|TRAP|Coated vesicle-associated kinase of 90 kDa|Teratoma-associated tyrosine kinase|TEIF|CVAK90|TAPK|N-terminal kinase-like protein|GKLP|Telomerase transcriptional element-interacting factor|Telomerase regulation-associated protein|SCY1-like protein 1
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 460-740 of human SCYL1 (NP_065731.3)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SCYL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat