Anti-IL11RA Rabbit Polyclonal Antibody
Catalog # PRSI22-362
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 11 receptor, alpha
- Antigen symbol:IL11RA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:3590
- Antigen synonyms:MGC2146|IL11RA|CRSDA
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 30-290 of human IL11RA (NP_001136256.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: IL11RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat