Anti-GTF2IRD1 Rabbit Polyclonal Antibody
PRSI22-351
New Product
- Antibody type:Primary
- Antigen name:GTF2I repeat domain containing 1
- Antigen symbol:GTF2IRD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:9569
- Antigen synonyms:RBAP2|hMusTRD1alpha1|WBSCR11|WBS|CREAM1|GTF3|MUSTRD1|GTF2IRD1|BEN|WBSCR12
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 660-959 of human GTF2IRD1 (NP_057412.1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Type: Primary
Antigen: GTF2IRD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat