Anti-SLC17A7 Rabbit Polyclonal Antibody
Catalog # PRSI16-881
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 17 member 7
- Antigen symbol:SLC17A7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:57030
- Antigen synonyms:BNPI|member 7|solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter)|brain-specific Na(+)-dependent inorganic phosphate cotransporter|vesicular glutamate transporter 1|solute carrier family 17 (vesicular glutamate transporter)|brain-specific Na-dependent inorganic phosphate cotransporter|VGLUT1
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 461-560 of human SLC17A7 (NP_064705,1)
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SLC17A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat