Anti-TADA3 Rabbit Polyclonal Antibody
Catalog # PRSI22-063
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Transcriptional adaptor 3
- Antigen symbol:TADA3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:10474
- Antigen synonyms:ADA3|TADA3L|hADA3|FLJ20221|STAF54|NGG1|FLJ21329
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 270-369 of human TADA3 (NP_597814.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: TADA3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat