Anti-RUVBL1 Rabbit Polyclonal Antibody
Catalog # PRSI19-815
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:RuvB-like AAA ATPase 1
- Antigen symbol:RUVBL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:8607
- Antigen synonyms:PONTIN|TIH1|NMP238|ECP54|TIP49A|Pontin52|RVB1|RUVBL1|TIP49|INO80H
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human RUVBL1 (NP_003698.1).
- Purification:Affinity purification
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: RUVBL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse