Anti-LCP1 Rabbit Polyclonal Antibody
Catalog # PRSI60-157
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Plastin-2
- Antigen symbol:LCP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:306 - 335 aa
- Form:Liquid
- Gene ID:3936
- Modification:None
- Molecular weight:70 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,35 mg/ml
- Shipping temperature:4°C
- Immunogen:This LCP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 306 - 335 amino acids from the Central region of human LCP1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: LCP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 306-335
Host: Rabbit
Isotype: Ig
Reactivity: Human