Anti-MED9 Rabbit Polyclonal Antibody
PRSI59-838
New Product
- Antibody type:Primary
- Antigen name:Mediator of RNA polymerase II transcription subunit 9
- Antigen symbol:MED9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:107 - 136 aa
- Form:Liquid
- Gene ID:55090
- Modification:None
- Molecular weight:16 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This MED9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 107 - 136 amino acids from the C-terminal region of human MED9.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: MED9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 107-136
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse