Anti-AGPAT9 Rabbit Polyclonal Antibody
Catalog # PRSI59-592
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Glycerol-3-phosphate acyltransferase 3
- Antigen symbol:AGPAT9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:194 - 222 aa
- Form:Liquid
- Gene ID:84803
- Modification:None
- Molecular weight:49 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4°C
- Immunogen:This AGPAT9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 194 - 222 amino acids from the Central region of human AGPAT9.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: AGPAT9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Amino Acid Range = 194-222
Host: Rabbit
Isotype: Ig
Reactivity: Human, Mouse