Order Entry
Finland
ContactUsLinkComponent
Human Recombinant ADAM-like decysin-1 (from Cells)
Human Recombinant ADAM-like decysin-1 (from Cells)
Catalog # PRSI91-838
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant ADAM-like decysin-1 (from Cells)
Catalog # PRSI91-838
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimise freeze-thaw cycles.
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Protein synonyms:
    a disintegrin and metalloproteinase domain-like protein decysin-1|ADAM-like protein decysin-1|ADAMDEC1|ADAM DEC1
  • Protein/peptide name:
    ADAM-like decysin-1
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    50,4 kDa
  • Sequence:
    Ile31-Glu470
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM Tris HCl, 150 mM NaCl, 1 mM ZnCl₂, pH 7,5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the Lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

ADAM DEC1 protein is expressed highly in the small intestine and appendix, moderately in lymph node, mucosal lining of the colon, thymus, spleen and very weakly in the bone marrow. ADAM DEC1 is induced during DC maturation and up-regulated in response to T-cell signals. It may play an important role in the control of the immune response and during pregnancy.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays.

Caution: For research use only.