Order Entry
Finland
ContactUsLinkComponent
Recombinant Protein G (from E. coli)
Recombinant Protein G (from E. coli)
Catalog # PRSI96-655
Supplier:  ProSci Inc.
CAS Number:  
undefined
Recombinant Protein G (from E. coli)
Catalog # PRSI96-655
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    2 mg
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Tag sequence:
    His Tag
  • Protein synonyms:
    RPG|Protein G
  • Protein/peptide name:
    Protein G
  • Purity:
    >98% as determined by SDS-PAGE
  • Formulation:
    Lyophilised, PBS, pH 7,4
  • Tested applications:
    Western blot

Specifications

About this item

Protein G is an immunoglobulin-binding protein expressed in group C and G Streptococcal bacteria much like Protein A but with differing specificities. It is a 65-kDa (G148 protein G) and a 58 kDa (C40 protein G) cell surface protein that has found application in purifying antibodies through its binding to the Fc region. The native molecule also binds albumin, however, because serum albumin is a major contaminant of antibody sources, the albumin binding site has been removed from this recombinant form of Protein G.