Anti-CPN1 Rabbit Polyclonal Antibody
PRSI56-714
New Product
- Antibody type:Primary
- Antigen name:Carboxypeptidase N catalytic chain
- Antigen symbol:CPN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Amino acid range = 210-240
- Form:Liquid
- Gene ID:P15169
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:52 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:2 mg/ml
- Shipping temperature:4 °C
- Immunogen:This CPN1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 210-240 amino acids from the Central region of human CPN1
- Purification:This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis
- Pk:400 µl
Carboxypeptidase N is a plasma metallo-protease that cleaves basic amino acids from the C terminal of peptides and proteins. The enzyme is important in the regulation of peptides like kinins and anaphylatoxins, and has also been known as kininase-1 and anaphylatoxin inactivator. This enzyme is a tetramer comprised of two identical regulatory subunits and two identical catalytic subunits; this gene encodes the catalytic subunit. Mutations in this gene can be associated with angioedema or chronic urticaria resulting from carboxypeptidase N deficiency. [provided by RefSeq].
- For WB starting dilution is: 1:1000
- For IHC-P starting dilution is: 1:10~50
- For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: CPN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human