Anti-PVRL3 Rabbit Polyclonal Antibody
Catalog # PRSI56-704
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Nectin-3
- Antigen symbol:PVRL3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino acid range = 507-536
- Form:Liquid
- Gene ID:Q9NQS3
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:61 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,47 mg/ml
- Shipping temperature:4 °C
- Immunogen:This PVRL3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 507-536 amino acids from the C-terminal region of human PVRL3
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
Nectins (e.g., PVRL1; MIM 600644) are immunoglobulin-like adhesion molecules that interact with afadin (AF6; MIM 159559). Afadin is an actin filament-binding protein that connects nectins to the actin cytoskeleton. The nectin-afadin system organizes adherens junctions cooperatively with the cadherin (see MIM 192090)-catenin (see MIM 116805) system in epithelial cells.
- For WB starting dilution is: 1:1000
Type: Primary
Antigen: PVRL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human