Anti-NUP35 Rabbit Polyclonal Antibody
Catalog # PRSI56-703
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:Nucleoporin NUP53
- Antigen symbol:NUP35
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- Western blot:Yes
- Epitope:Amino acid range = 289-318
- Form:Liquid
- Gene ID:Q8NFH5
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:35 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This NUP35 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 289-318 amino acids from the C-terminal region of human NUP35
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification
- Pk:400 µl
Specifications
About this item
This gene encodes a member of the nucleoporin family. The protein is localized to the nuclear rim and is part of the nuclear pore complex (NPC). All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC.
- For WB starting dilution is: 1:1000
- For FACS starting dilution is: 1:10~50
Type: Primary
Antigen: NUP35
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human