Order Entry
Finland
ContactUsLinkComponent
Anti-Cd8a Rat Monoclonal Antibody [clone: 2.43] (VioletFluor® 450)
Anti-Cd8a Rat Monoclonal Antibody [clone: 2.43] (VioletFluor® 450)
Catalog # PRSI76-648
Supplier:  ProSci Inc.
New Product
Anti-Cd8a Rat Monoclonal Antibody [clone: 2.43] (VioletFluor® 450)
Catalog # PRSI76-648
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD8 antigen, alpha chain
  • Antigen symbol:
    Cd8a
  • Clonality:
    Monoclonal
  • Clone:
    2.43
  • Conjugation:
    VioletFluor® 450
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01731
  • Antigen synonyms:
    Cd8a
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

The 2.43 monoclonal antibody specifically reacts with mouse CD8 antigen. CD8a (the alpha chains) form heterodimers with CD8b (the beta chains) or homodimers (alpha-alpha), which occur as receptors on the surface of the majority of thymocytes. A subpopulation of mature T lymphocytes expresses the CD8 alpha beta (alpha beta TCR T cells), and a subpopulation of intestinal intraepithelial lymphocytes and dendritic cells express CD8a without CD8b. CD8 interacts with the mouse major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450 nm.

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: 2.43
Conjugation: VioletFluor® 450
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse