Order Entry
Finland
ContactUsLinkComponent
Anti-TNF Mouse Monoclonal Antibody [clone: MAb11] (PE (Phycoerythrin))
Anti-TNF Mouse Monoclonal Antibody [clone: MAb11] (PE (Phycoerythrin))
Catalog # PRSI77-183
Supplier:  ProSci Inc.
New Product
Anti-TNF Mouse Monoclonal Antibody [clone: MAb11] (PE (Phycoerythrin))
Catalog # PRSI77-183
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    tumor necrosis factor
  • Antigen symbol:
    TNF
  • Clonality:
    Monoclonal
  • Clone:
    MAb11
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01375
  • Antigen synonyms:
    TNF
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    100 Tests

Specifications

About this item

The MAb11 monoclonal antibody specifically reacts with human tumor necrosis factor alpha (TNF-alpha), a 157 amino acid non-glycosylated protein. TNF-alpha is a pleiotropic pro-inflammatory cytokine secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. It belongs to the TNF family of ligands and signals through two receptors, TNFR1 and TNFR2. TNF-alpha is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-alpha also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. The MAB11 antibody was generated from recombinant human TNF and is reported to be a neutralizing antibody.

Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: MAb11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human