Order Entry
Finland
ContactUsLinkComponent
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: LN3] (APC (Allophycocyanin))
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: LN3] (APC (Allophycocyanin))
Catalog # PRSI77-014
Supplier:  ProSci Inc.
New Product
Anti-HLA-DRA Mouse Monoclonal Antibody [clone: LN3] (APC (Allophycocyanin))
Catalog # PRSI77-014
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    major histocompatibility complex, class II, DR alpha
  • Antigen symbol:
    HLA-DRA
  • Clonality:
    Monoclonal
  • Clone:
    LN3
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P01903
  • Antigen synonyms:
    HLA-DRA
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    100 Tests

Specifications

About this item

The LN3 monoclonal antibody specifically reacts with the human leukocyte antigen-DR (HLA-DR), a MHC class II heterodimeric surface glycoprotein. HLA-DR is expressed on the antigen presenting surfaces of monocytes, macrophages, dendritic cells, activated T cells, and B cells. It is integral to peptide presentation to CD4+ T cells.

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: LN3
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human