Order Entry
Finland
ContactUsLinkComponent
Anti-Foxp3 Mouse Monoclonal Antibody [clone: 3G3] (PE (Phycoerythrin))
Anti-Foxp3 Mouse Monoclonal Antibody [clone: 3G3] (PE (Phycoerythrin))
Catalog # PRSI77-149
Supplier:  ProSci Inc.
New Product
Anti-Foxp3 Mouse Monoclonal Antibody [clone: 3G3] (PE (Phycoerythrin))
Catalog # PRSI77-149
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    forkhead box P3
  • Antigen symbol:
    Foxp3
  • Clonality:
    Monoclonal
  • Clone:
    3G3
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    Q99JB6
  • Antigen synonyms:
    Foxp3
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,025 mg

Specifications

About this item

Type: Primary
Antigen: Foxp3
Clonality: Monoclonal
Clone: 3G3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse