Anti-Itgb1 Hamster Monoclonal Antibody [clone: HMb1-1] (PE (Phycoerythrin))
Catalog # PRSI76-798
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:integrin beta 1 (fibronectin receptor beta)
- Antigen symbol:Itgb1
- Clonality:Monoclonal
- Clone:HMb1-1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:P09055
- Antigen synonyms:Itgb1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,05 mg
Specifications
About this item
The HMb1-1 monoclonal antibody specifically reacts with mouse/rat CD29, a 130 kDa molecule also known as integrin beta 1, GPIIa, and the VLA-beta chain. It is expressed broadly on endothelial cells, epithelial, leukocytes, and smooth muscle. CD29 forms the VLA 1-6 complexes through the non-covalent interaction with the alpha integrins of CD49 a-f. The HMb1-1 antibody is capable of inhibiting T cell proliferation and blocking cell adhesion.
Type: Primary
Antigen: Itgb1
Clonality: Monoclonal
Clone: HMb1-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse;Rat