Anti-ITGB1 Mouse Monoclonal Antibody [clone: TS2/16] (FITC (Fluorescein Isothiocyanate))
Catalog # PRSI76-790
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includes MDF2, MSK12)
- Antigen symbol:ITGB1
- Clonality:Monoclonal
- Clone:TS2/16
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P05556
- Antigen synonyms:ITGB1
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:100 Tests
Specifications
About this item
The TS2/16 moncolonal antibody specifically reacts with human CD29 (Integrin beta1), a 130 kDa type I glycoprotein expressed on hematopoietic and non-hematopoietic cells. It forms the VLA-(1-6) molecules with integrin alpha-(1-6) and is involved in the adhesion between cell-cell and cell-matrix. The TS2/16 antibody has been found to activate beta 1 integrins.
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: TS2/16
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human