Order Entry
Finland
ContactUsLinkComponent
Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (PE (Phycoerythrin)-Cy7®)
Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (PE (Phycoerythrin)-Cy7®)
Catalog # PRSI76-545
Supplier:  ProSci Inc.
New Product
Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (PE (Phycoerythrin)-Cy7®)
Catalog # PRSI76-545
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    protein tyrosine phosphatase, receptor type, C
  • Antigen symbol:
    Ptprc
  • Clonality:
    Monoclonal
  • Clone:
    A20
  • Conjugation:
    PE (Phycoerythrin)-Cy7®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    P06800
  • Antigen synonyms:
    Ptprc
  • Storage buffer:
    Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
  • Shipping temperature:
    4 °C
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: A20
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse