Order Entry
Finland
ContactUsLinkComponent
 
Anti-CD48 Mouse Monoclonal Antibody [clone: 156-4H9]
Catalog # PRSI76-756
Supplier:  ProSci Inc.
New Product
Anti-CD48 Mouse Monoclonal Antibody [clone: 156-4H9]
Catalog # PRSI76-756
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD48 molecule
  • Antigen symbol:
    CD48
  • Clonality:
    Monoclonal
  • Clone:
    156-4H9
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    purified antibody
  • Form:
    liquid
  • Gene ID:
    P09326
  • Antigen synonyms:
    CD48
  • Storage buffer:
    Phosphate-buffered aqueous solution, ph 7,2.
  • Storage temperature:
    The product should be stored undiluted at 4 °C. Do not freeze.
  • Shipping temperature:
    4 °C
  • Tested applications:
    Functional
  • Purification:
    The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0,01 EU/µg of the protein.
  • Pk:
    0,05 mg

Specifications

About this item

The 156-4H9 monoclonal antibody specifically reacts with human CD48, a GPI-linked glycoprotein in the Ig superfamily. CD48 was previously known as BCM1 in mice, Blast-1 in humans, and OX-45 in rats. Its ligands are CD2 and CD244 and it is expressed on leukocytes to the exclusion of non-hematopoietic cells and its expression increases in B cells upon activation.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 156-4H9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human