Anti-Ptprc Mouse Monoclonal Antibody [clone: A20] (PE (Phycoerythrin))
Catalog # PRSI76-541
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:protein tyrosine phosphatase, receptor type, C
- Antigen symbol:Ptprc
- Clonality:Monoclonal
- Clone:A20
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Cross adsorption:No
- Form:liquid
- Gene ID:P06800
- Antigen synonyms:Ptprc
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: Ptprc
Clonality: Monoclonal
Clone: A20
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse