Anti-PTPRC Mouse Monoclonal Antibody [clone: HI100] (VioletFluor® 450)
Catalog # PRSI76-456
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:protein tyrosine phosphatase, receptor type, C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:HI100
- Conjugation:VioletFluor® 450
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:P08575
- Antigen synonyms:PTPRC
- Storage buffer:Phosphate-buffered aqueous solution, ≤0,09% Sodium azide, may contain carrier protein/stabiliser, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C and should be protected from prolonged exposure to light. Do not freeze.
- Shipping temperature:4 °C
- Purification:The monoclonal antibody was purified utilizing affinity chromatography and unreacted dye was removed from the product.
- Pk:25 Tests
Specifications
About this item
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI100
Conjugation: VioletFluor® 450
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human