Anti-Cd27 Hamster Monoclonal Antibody [clone: LG.3A10]
Catalog # PRSI76-630
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD27 antigen
- Antigen symbol:Cd27
- Clonality:Monoclonal
- Clone:LG.3A10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Hamster
- Isotype:IgG
- Reactivity:Human,Mouse
- Cross adsorption:No
- Format:purified antibody
- Form:liquid
- Gene ID:P41272
- Antigen synonyms:Cd27
- Storage buffer:Phosphate-buffered aqueous solution, ph 7,2.
- Storage temperature:The product should be stored undiluted at 4 °C. Do not freeze.
- Shipping temperature:4 °C
- Tested applications:Functional
- Purification:The monoclonal antibody was purified utilizing affinity chromatography. The endotoxin level is determined by LAL test to be less than 0,01 EU/µg of the protein.
- Pk:0,05 mg
Specifications
About this item
The LG.3A10 monoclonal antibody binds to CD27, 45 kDa molecule in the Tumor Necrosis Factor Receptor (TNFR) superfamily. It is expressed on memory B cells, NK cells, mature T cells, and a subset of thymocytes. Cd27 is involved in the interactions between B and T cells through its ligand CD70. The LG.3A10 antibody is cross-reactive with human, mouse and rat CD27.
Type: Primary
Antigen: Cd27
Clonality: Monoclonal
Clone: LG.3A10
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Human;Mouse