Anti-Atp1a1 Rabbit Polyclonal Antibody
PRSI61-010
- Antibody type:Primary
- Antigen name:Sodium/potassium-transporting ATPase subunit alpha-1
- Antigen symbol:Atp1a1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:24211
- Modification:None
- Molecular weight:113 kDa
- Storage temperature:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Shipping temperature:4 °C
- Immunogen:This Rat Atp1a1 antibody is generated from a rabbit immunised with a KLH conjugated synthetic peptide between 1 - 30 amino acids from Rat Atp1a1.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Pk:400 µl
For WB starting dilution is: 1:1000.
Type: Primary
Antigen: Atp1a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Ig
Reactivity: Rat