Thymus Lysate (7 Days Old)
Catalog # PRSI1409-7
Supplier: ProSci Inc.
New Product
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Pk:0,1 mg
- Description:Normal; Source: Mouse; Tissue: Thymus
- Storage conditions:Store at 2...8 °C for continuous use. For extended storage, freeze working aliquots at −70 °C.
- Storage buffer:Thymus lysate (7 Days Old) is supplied in SDS sample buffer containing 5% β-mercaptoethanol
- Concentration:2 mg/ml
- Tested applications:WB
Specifications
About this item
Thymus tissue lysate (7 Days Old) was prepared by homogenisation in modified RIPA buffer (150 mM sodium chloride, 50 mM Tris-HCl, pH 7,4, 1 mM ethylenediaminetetraacetic acid, 1 mM phenylmethylsulfonyl fluoride, 1% Triton X-100, 1% sodium deoxycholic acid, 0.1% sodium dodecylsulfate, 5 μg/ml of aprotinin, 5 μg/ml of leupeptin.
Tissue and cell debris was removed by centrifugation. Protein concentration was determined with Bio-Rad protein assay. The product was boiled for 5 min in 1X SDS sample buffer (50 mM Tris-HCl pH 6,8, 12,5% glycerol, 1% sodium dodecylsulfate, 0,01% bromophenol blue) containing 5% β-mercaptoethanol.