Anti-EIF3J Rabbit Polyclonal Antibody
Catalog # PRSI23-694
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:eukaryotic translation initiation factor 3 subunit J
- Antigen symbol:EIF3J
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Form:Liquid
- Gene ID:8669
- Antigen synonyms:subunit 1 (alpha|eIF3-alpha|eukaryotic translation initiation factor 3|EIF3S1|eIF3-p35|eukaryotic translation initiation factor 3 subunit J|35kD)
- Modification:None
- Storage buffer:PBS with 0,02% sodium azide, 50% glycerol, pH 7,3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-258 of human EIF3J (NP_003749.2)
- Purification:Affinity purification.
- Pack type:Vial
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: EIF3J
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat