Anti-Havcr1 Recombinant Antibody [Clone: 3B3]
Catalog # PRSI24-600
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:hepatitis A virus cellular receptor 1
- Antigen symbol:Havcr1
- Clonality:Recombinant
- Clone:3B3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Cross adsorption:No
- Form:Liquid
- Gene ID:171283
- Antigen synonyms:TIM1|TIM-1|KIM-1|CD365|T-cell immunoglobulin mucin receptor 1|T cell immunoglobulin and mucin domain-containing protein 1|HAVCR1|T-cell immunoglobulin and mucin domain containing 1|Kidney injury molecule 1|HAVcr-1|Hepatitis A virus cellular receptor 1 homolog|T cell membrane protein 1|TIMD-1
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:This antibody was raised by immunising female Lewis strain rats (Harlan Sprague-Dawley) subcutaneously with mouse TIM-1–Ig in complete Freund's adjuvant (CFA), followed by multiple 'boosting' with mouse TIM-1–Ig in PBS
- Tested applications:Agonism
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: hepatitis A virus cellular receptor 1
Clonality: Recombinant
Clone: 3B3
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2a kappa
Reactivity: Mouse