Anti-CLEC4M Recombinant Antibody [Clone: 16E7]
Catalog # PRSI24-134
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Antigen name:HGNC:HGNC:13523
- Antigen symbol:CLEC4M
- Clonality:Recombinant
- Clone:16E7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Cross adsorption:No
- Form:Liquid
- Gene ID:10332
- Antigen synonyms:C-type lectin domain family 4 member M|DC-SIGN-related protein|DC-SIGN2|Dendritic cell-specific ICAM-3-grabbing non-integrin 2|CD299|CLEC4M|CD209 antigen-like protein 1|Liver/lymph node-specific ICAM-3-grabbing non-integrin|L-SIGN
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:CD29
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: HGNC:HGNC:13523
Clonality: Recombinant
Clone: 16E7
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG kappa
Reactivity: Human