Anti-NIP Recombinant Antibody [Clone: B1-8]
Catalog # PRSI24-098
Supplier: ProSci Inc.
New Product
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:B1-8
- Conjugation:Unconjugated
- Isotype:IgG lambda
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:NIP
- Modification:Unmodified
- Storage buffer:PBS with 0,02% Proclin 300
- Storage temperature:Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
- Shipping temperature:Shipped on blue ice
- Immunogen:hapten-protein conjugated NP12-chicken gammaglobulin (CG)
- Tested applications:Negative control
- Pack type:Vial
- Pk:0,2 mg
Specifications
About this item
Type: Primary
Antigen: NIP
Clonality: Recombinant
Clone: B1-8
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG lambda
Reactivity: