Order Entry
Finland
ContactUsLinkComponent
Anti-His tag Recombinant Antibody [Clone: 3D5]
Anti-His tag Recombinant Antibody [Clone: 3D5]
Catalog # PRSI24-057
Supplier:  ProSci Inc.
New Product
Anti-His tag Recombinant Antibody [Clone: 3D5]
Catalog # PRSI24-057
Supplier:  ProSci Inc.
New Product

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Recombinant
  • Clone:
    3D5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Isotype:
    IgG2 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    polyhistidine tag
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Purified his tagged Fab fragment
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: His tag
Clonality: Recombinant
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2 kappa
Reactivity: