Order Entry
Finland
ContactUsLinkComponent
 
Acid hematoxylin solution
  GHS280-2.5L
 :  
Acid hematoxylin solution
  GHS280-2.5L
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096

 

 

  • Appearance (Turbidity):
    Clear
  • Appearance(Colour)
    Reddish-Purple:
    Conforms
  • Appearance (Form)
    Essentially free of precipitate:
    Solution
  • Performance:
    Conforms
  • Expiration Date Period:
    24 months

 

 

 :
 :M-Clarity™ quality level = MQ500 (more info.)
 :GHS05
 :Room Temperature