Starch 1% in water stabilised indicator, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:9005-25-8
:
Formula:(C₆H₁₀O₅)n
MDL Number:MFCD00082026
Density:1.005 g/cm³
:Stabilised with mercury (II) iodide
:162.14 g/mol
Storage Temperature:Ambient
EINECS:232-679-6
Specification Test Results
- Turbidity:Max. 50 NTU
- Test of sensitivity:complying