Devarda's alloy, Supelco®
Catalog # 31385-250G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
- Seq:3545
Specifications
About this item
:
Merck Index:13,02958
Boiling Pt:906 °C (1013 hPa)
Melting Pt:490…560 °C
Density:5.79 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:3089
ADR:4.1,II
:M-Clarity™ quality level = MQ100
CAS Number:8049-11-4
MDL Number:MFCD00198176