Toluidine blue, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:6586-04-5
:
Formula:C₁₅H₁₆ClN₃S·0.5ZnCl₂
MDL Number:MFCD03548293
:M-Clarity™ quality level = MQ200
:373.97 g/mol
Storage Temperature:Room Temperature
Specification Test Results
- Grade:for microscopy (Hist., Vit.)
- Quality level:200
- Solubility:Methanol: 0,01 g/10 ml, blue to very deep blue
- εmax:≥800 at 621 - 636 nm in water